Key Documents
MDFNSPF-L
Mediflex Nisense Gloves, blue nitrile, powder free
size L
About This Item
UNSPSC Code:
46181504
Pricing and availability is not currently available.
material
sky blue nitrile
size
L
description
textured
feature
latex free
powder-free
packaging
box of 100 gloves
L
240 mm , cuff
General description
Powder free blue nitrile gloves. Non sterile, ambidextrous, latex
free, ultra thin, 240mm cuff, textured. Single use only.
Choose from one of the most recent versions:
Certificates of Analysis (COA)