Skip to Content
MilliporeSigma
All Photos(1)

Documents

M78607

Sigma-Aldrich

6-Methyl-2-pyridinemethanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO
CAS Number:
Molecular Weight:
123.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

105-108 °C/12 mmHg (lit.)

mp

32-34 °C (lit.)

SMILES string

Cc1cccc(CO)n1

InChI

1S/C7H9NO/c1-6-3-2-4-7(5-9)8-6/h2-4,9H,5H2,1H3

InChI key

JLVBSBMJQUMAMW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salah S Massoud et al.
Dalton transactions (Cambridge, England : 2003), 43(26), 10086-10103 (2014-05-30)
Four new cobalt(ii) complexes [Co(6-MeTPA)Cl]ClO4/PF6 (2/2a), [Co(6-Me2TPA)Cl]ClO4/PF6 (3/3a), [Co(BPQA)Cl]ClO4/PF6 (4/4a) and [Co(BQPA)Cl]ClO4/PF6 (5/5a) as well as [Co(TPA)Cl]ClO4 (1) where TPA = tris(2-pyridylmethyl)amine, 6-MeTPA = ((6-methyl-2-pyridyl)methyl)bis(2-pyridylmethyl)amine, 6-Me2TPA = bis(6-methyl-2-pyridyl)methyl)-(2-pyridylmethyl)amine, BPQA = bis(2-pyridylmethyl)-(2-quinolylmethyl)-amine and BQPA = bis(2-quinolylmethyl)-(2-pyridylmethyl)amine were synthesized and structurally characterized.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service