Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M73001

Sigma-Aldrich

3-Methylpiperidine

99%

Synonym(s):

β-Pipecoline, 3-Pipecoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
Beilstein:
79807
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.447 (lit.)

bp

125-126 °C/763 mmHg (lit.)

density

0.845 g/mL at 25 °C (lit.)

SMILES string

CC1CCCNC1

InChI

1S/C6H13N/c1-6-3-2-4-7-5-6/h6-7H,2-5H2,1H3

InChI key

JEGMWWXJUXDNJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of:
  • Phenylpropenamide derivatives for anti-hepatitis B virus activity
  • CB2 receptor agonists for treatment of chronic pain
  • Aurora kinase inhibitors
  • Spiroimidazolidinone NPC1L1 inhibitors
  • 1,3,5-Oxadiazinones
  • Selective serotonin 5-HT6 receptor antagonists

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service