Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M67109

Sigma-Aldrich

4-Methyl-2-pentanone

≥99%

Synonym(s):

Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCH3
CAS Number:
Molecular Weight:
100.16
Beilstein:
605399
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

vapor density

3.5 (vs air)

vapor pressure

15.7 mmHg

Assay

≥99%

autoignition temp.

840 °F

expl. lim.

1.2-8 %, 93 °F

refractive index

n20/D 1.395 (lit.)

bp

117-118 °C
117-118 °C

mp

−80 °C (lit.)

density

0.801 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(C)=O

InChI

1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3

InChI key

NTIZESTWPVYFNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H J Shim et al.
Journal of chromatography. B, Biomedical applications, 656(2), 407-414 (1994-06-17)
A high-performance liquid chromatographic method was developed for the simultaneous determination of a new anthracycline, DA-125 (I), and its metabolites (M1, M2, M3, and M4) in rat plasma and urine using fluorescein as an internal standard. Compound I, a prodrug
Ginette Truchon et al.
Journal of occupational and environmental hygiene, 3(3), 137-143 (2006-02-09)
Compartmental and physiologically based toxicokinetic modeling coupled with Monte Carlo simulation were used to quantify the impact of biological variability (physiological, biochemical, and anatomic parameters) on the values of a series of bio-indicators of metal and organic industrial chemical exposures.
Vincent W Chen et al.
Optics express, 22(10), 12316-12326 (2014-06-13)
Cuboid-shaped organic microcavities containing a pyrromethene laser dye and supported upon a photonic crystal have been investigated as an approach to reducing the lasing threshold of the cavities. Multiphoton lithography facilitated fabrication of the cuboid cavities directly on the substrate
David Quesnel et al.
Water research, 39(4), 677-687 (2005-02-15)
An industrial wastewater that was pretreated by an aerobic thermophilic bacterial consortium (THE) was subjected to additional treatability studies by granular activated carbon (GAC) and a conventional activated sludge (CAS). The removal of dissolved organic carbon (DOC) in both systems
Mao Ting et al.
The Science of the total environment, 390(1), 97-108 (2007-11-03)
Volatile organic compounds (VOCs) in the air of Beijing City were measured at the heights of 8, 32, 140 and 280 m on the Beijing 325 m meteorological tower in autumn 2005. Concentrations of fifty-five compounds were determined by quantitative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service