Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M59807

Sigma-Aldrich

4-Methyl-3-nitroaniline

97%

Synonym(s):

3-Nitro-4-methylaniline, 3-Nitro-p-toluidine, 4-Amino-2-nitro-toluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)NH2
CAS Number:
Molecular Weight:
152.15
Beilstein:
638984
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

74-77 °C (lit.)

SMILES string

Cc1ccc(N)cc1[N+]([O-])=O

InChI

1S/C7H8N2O2/c1-5-2-3-6(8)4-7(5)9(10)11/h2-4H,8H2,1H3

InChI key

GDIIPKWHAQGCJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seok-Young Oh et al.
Bioresource technology, 216, 1014-1021 (2016-06-28)
In order to remediate organic contaminants in natural waters and soils, a novel zero-valent iron [Fe(0)]-included biochar was synthesized via slow pyrolysis. 2,4-Dinitrotoluene (DNT) and 2,4-dichlorophenol (DCP) were removed in water via sorption to the Fe(0)-included biochar. Compared to sorption
P Z Lang et al.
Chemosphere, 35(8), 1799-1815 (1997-11-14)
Bioconcentration curves of 2,4-dinitrotoluene(2,4-DNT) in carps (whole fish, liver, intestine and muscle) were investigated using semistatic system. For whole fish, its curve could be described as a gentle peak which began with a rise in concentration to summit or steady
J A Bond et al.
Drug metabolism and disposition: the biological fate of chemicals, 9(1), 10-14 (1981-01-01)
Fischer-344 rat hepatocytes display a capacity for both oxidation and reduction of 2,4-dinitrotoluene (2,4-DNT), a potent hepatocarcinogen. The major metabolite detected by high-pressure liquid chromatography was 2,4-dinitrobenzyl alcohol (2,4-DNBalc), which accounted for 75-80% of the total metabolites formed. The apparent
Z-Y Wang et al.
Journal of applied microbiology, 110(6), 1476-1484 (2011-03-15)
To evaluate the biodegradability of 2,4-DNT using an anaerobic filter (AF) combined with a biological aerated filter (BAF), and elucidate the degradation mechanism of 2,4-DNT and analyze the bacterial community of the reactors over a long period of operation. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service