Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M3809

Sigma-Aldrich

2-Mercaptoethyl ether

95%

Synonym(s):

2,2′-Oxydiethanethiol, Bis(2-mercaptoethyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(CH2CH2SH)2
CAS Number:
Molecular Weight:
138.25
Beilstein:
969211
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

bp

217 °C (lit.)

mp

−80 °C (lit.)

density

1.114 g/mL at 25 °C (lit.)

SMILES string

SCCOCCS

InChI

1S/C4H10OS2/c6-3-1-5-2-4-7/h6-7H,1-4H2

InChI key

CNDCQWGRLNGNNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prarthana Patil et al.
Tissue engineering. Part C, Methods, 23(11), 754-762 (2017-08-02)
Impaired wound healing that mimics chronic human skin pathologies is difficult to achieve in current animal models, hindering testing and development of new therapeutic biomaterials that promote wound healing. In this article, we describe a refinement and simplification of the
Nana Lu et al.
Biomacromolecules, 20(9), 3485-3493 (2019-07-30)
Organic-inorganic oligo(ethylene glycol)-polyhedral oligomeric silsesquioxanes (OEG n
Zuoti Xie et al.
ACS nano, 11(1), 569-578 (2016-12-10)
The transport properties of molecular junctions based on alkanedithiols with three different methylene chain lengths were compared with junctions based on similar chains wherein every third -CH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service