Skip to Content
MilliporeSigma
All Photos(1)

Documents

M2509

Sigma-Aldrich

3-Methylxanthine

Synonym(s):

2,6-Dihydroxy-3-methylpurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O2
CAS Number:
Molecular Weight:
166.14
Beilstein:
180944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

mp

>300 °C (lit.)

storage temp.

2-8°C

SMILES string

CN1C(=O)NC(=O)c2[nH]cnc12

InChI

1S/C6H6N4O2/c1-10-4-3(7-2-8-4)5(11)9-6(10)12/h2H,1H3,(H,7,8)(H,9,11,12)

InChI key

GMSNIKWWOQHZGF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Metabolite of theophylline that is pharmacologically active but less potent than theophylline.

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Tanaka et al.
Clinical and experimental pharmacology & physiology, 18(3), 163-168 (1991-03-01)
1. For the purpose of clarifying the mechanism of the airways smooth muscle relaxant action of xanthines, cyclic guanosine monophosphate (GMP) phosphodiesterase (PDE) from guinea-pig trachealis muscle was purified with diethylaminoethyl ether (DEAE) cellulose column chromatography. 2. Five 3-alkylxanthines (3-methylxanthine
K Yamamoto et al.
Biological & pharmaceutical bulletin, 19(6), 869-872 (1996-06-01)
To evaluate the risk of neurotoxicity induced by theophylline and its main metabolites, 1-methylxanthine (1-MX), 3-methylxanthine (3-MX), 1,3-dimethyluric acid (1,3-DMUA) and 1-methyluric acid (1-MUA), we compared their convulsive potency to central nervous system (CNS) after intracerebral administration to mice. All
H R Ha et al.
British journal of clinical pharmacology, 39(3), 321-326 (1995-03-01)
1. Theophylline metabolism was studied using seven human cytochrome P-450 isoforms (CYPs), namely CYP1A1, 1A2, 2A6, 2B6, 2D6, 2E1 and 3A4, and microsomal epoxide hydroxylase (EH), expressed in human B-lymphoblastoid cell lines. 2. At a high theophylline concentration of 10

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service