Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M23500

Sigma-Aldrich

3-(2-Methoxyphenyl)propionic acid

98%

Synonym(s):

2-Methoxyhydrocinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CH2CO2H
CAS Number:
Molecular Weight:
180.20
Beilstein:
2097012
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

85-87 °C (lit.)

SMILES string

COc1ccccc1CCC(O)=O

InChI

1S/C10H12O3/c1-13-9-5-3-2-4-8(9)6-7-10(11)12/h2-5H,6-7H2,1H3,(H,11,12)

InChI key

XSZSNLOPIWWFHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service