Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

L4001

Sigma-Aldrich

3,4-Lutidine

98%

Synonym(s):

3,4-Dimethylpyridine

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$100.00

$100.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$100.00

About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
Beilstein:
106583
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$100.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.511 (lit.)

bp

163-164 °C (lit.)

mp

−12 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

Cc1ccncc1C

InChI

1S/C7H9N/c1-6-3-4-8-5-7(6)2/h3-5H,1-2H3

InChI key

NURQLCJSMXZBPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Wleklinski et al.
Physical chemistry chemical physics : PCCP, 17(28), 18364-18373 (2015-06-25)
Electrospray ionization of metal salt solutions followed by ambient heating transforms the resulting salt clusters into new species, primarily naked ionic metal clusters. The experiment is done by passing the clusters through a heated coiled loop outside the mass spectrometer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service