Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L040050

Sigma-Aldrich

Cap A

configured for (ÄKTA® and OligoPilot®)

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
$723.00

$723.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2.5 L
$723.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.21

$723.00


Please contact Customer Service for Availability

Request a Bulk Order

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

form

liquid

storage condition

dry at room temperature

impurities

≤100 ppm water content (Karl Fischer)

color

clear

suitability

conforms to structure (GC, Formulation)

compatibility

configured for (ÄKTA® and OligoPilot®)

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Cap A solution is used after the phosphoramidite reaction/coupling step. This solution aborts chains bearing unreacted 5′hydroxyl groups due to failure of reaction with activated phosphoramidite (typically, 1 to 2% of growing oligonucleotide chains in each synthesis cycle). These unreacted 5′hydroxyl groups are capped with an acetyl group and thus rendered unreactive for subsequent synthesis steps.

Preparation Note

Acetonitrile / N-Methylimidazole 8/2 (v/v)

Other Notes

Cap A solution, containing acetonitrile (ACN) and N-methylimidazole (NMI), is mixed in situ with Cap B1 and Cap B2 solution during the capping (acetylation) reaction. Pyridine is applied as a mild base while NMI provides a powerful acylation catalyst.

Legal Information

OligoPilot is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service