Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JKA7077

Sigma-Aldrich

4arm-PEG10K 2arm-OH 2arm-NH2

HCl Salt, average Mn 10,000

Synonym(s):

4arm-PEG 2arm-OH 2arm-NH2

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$995.00

$995.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$995.00

About This Item

Linear Formula:
C(CH2O(CH2CH2O)nCH2CH2NH2HCl)2(CH2O(CH2CH2O)nH)2
UNSPSC Code:
12162002
NACRES:
NA.23

$995.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

mol wt

average Mn 10,000

reaction suitability

reactivity: carboxyl reactive

polymer architecture

shape: 4-arm
functionality: heterofunctional

storage temp.

−20°C

General description

Bifunctional multi-arm PEG reagent. It can connect two drugs at the same time. It is also useful for ADCs, increasing drug loading.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service