Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JKA13021

Sigma-Aldrich

mPEG7-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2CH2O)6CH2CH2NH2
UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

reaction suitability

reagent type: chemical modification reagent
reactivity: carboxyl reactive

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

General description

Monodisperse PEG. The amine group reacts with a carboxyl group (-COOH) and other amine reactive chemical groups.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service