Skip to Content
MilliporeSigma
All Photos(1)

Documents

H9605

Sigma-Aldrich

1,2,3,6,7,8-Hexahydropyrene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16
CAS Number:
Molecular Weight:
208.30
Beilstein:
2048996
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

130-134 °C (lit.)

SMILES string

C1Cc2ccc3CCCc4ccc(C1)c2c34

InChI

1S/C16H16/c1-3-11-7-9-13-5-2-6-14-10-8-12(4-1)15(11)16(13)14/h7-10H,1-6H2

InChI key

MBAIEZXRGAOPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Important intermediate for the synthesis of polycyclic hydrocarbons.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 111, 4285-4285 (1989)
A J Dobson et al.
Acta crystallographica. Section C, Crystal structure communications, 52 ( Pt 12), 3086-3088 (1996-12-15)
In 1,2,3,6,7,8-hexahydropyrene, C16H16, the unsaturated rings lie essentially in a plane, the r.m.s. deviation of the atoms defining this plane form the best-fit plane being 0.006 (2) A. The saturated rings are substantially non-planar, but the character of the non-planarity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service