Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H7637

Sigma-Aldrich

1-Hexadecanethiol

technical grade, 92%

Synonym(s):

Cetyl mercaptan, Hexadecyl mercaptan, Mercaptan C16

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15SH
CAS Number:
Molecular Weight:
258.51
Beilstein:
1748495
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

grade

technical grade

Assay

92%

refractive index

n20/D 1.462 (lit.)

bp

184-191 °C/7 mmHg (lit.)

mp

18-20 °C (lit.)

density

0.84 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCS

InChI

1S/C16H34S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3

InChI key

ORTRWBYBJVGVQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service