Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H61200

Sigma-Aldrich

Hypoxanthine

98%

Synonym(s):

6-Hydroxypurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4O
CAS Number:
Molecular Weight:
136.11
Beilstein:
5811
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

98%

mp

>300 °C (lit.)

SMILES string

O=C1NC=Nc2nc[nH]c12

InChI

1S/C5H4N4O/c10-5-3-4(7-1-6-3)8-2-9-5/h1-2H,(H2,6,7,8,9,10)

InChI key

FDGQSTZJBFJUBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hypoxanthine is a nutrient additive for a variety of cell culture applications involving bacterial, parasite (Plasmodium falciparum) and animal cells. Hypoxanthine is a component of selection media used in hybridoma technologies.

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John V W Becker et al.
Malaria journal, 10, 295-295 (2011-10-12)
Anti-malarial drug resistance threatens to undermine efforts to eliminate this deadly disease. The resulting omnipresent requirement for drugs with novel modes of action prompted a national consortium initiative to discover new anti-plasmodial agents from South African medicinal plants. One of
Tomáš Tichý et al.
European journal of medicinal chemistry, 55, 307-314 (2012-08-04)
Hexadecyloxypropyl esters of acyclic nucleoside phosphonates containing guanine (G) or hypoxanthine (Hx) and a (S)-[3-hydroxy-2-(phosphonomethoxy)propyl] [(S)-HPMP] or 2-(2-phosphonoethoxy)ethyl (PEE) acyclic moiety have been prepared. The activity of the prodrugs was evaluated in vitro against different virus families. Whereas ester derivatives of
Abdulazeez T Lawal et al.
Food chemistry, 135(4), 2982-2987 (2012-09-18)
A potentiometric enzyme electrode for detection of hypoxanthine (Hx) in fish meat is described. The sensor was developed by entrapment of xanthine oxidase (XOD) and ferrocene carboxylic acid (Fc) into polypyrrole (PPy) film during galvanostatic polymerisation film formation. The responses
Chien-Wei Hou et al.
The American journal of Chinese medicine, 40(5), 979-991 (2012-08-30)
Hyperuricemia causes gouty arthritis, kidney disease, heart disease, and other diseases. Xanthine oxidase (XOD) and urate transporters play important roles in urate homeostasis. Numerous plants have been identified as XOD inhibitors. Longan seeds are known to contain high levels of
Chia-Tung Wu et al.
Cardiovascular research, 102(3), 487-496 (2014-03-07)
Fibroblasts, which play an important role in cardiac function/dysfunction, including arrhythmogenesis, have voltage-dependent (Kv) currents of unknown importance. Here, we assessed the differential expression of Kv currents between atrial and ventricular fibroblasts from control dogs and dogs with an atrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service