Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H48204

Sigma-Aldrich

4-Hydroxy-3-nitrobenzenearsonic acid

98%

Synonym(s):

2-Nitrophenol-4-arsonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(NO2)AsO3H2
CAS Number:
Molecular Weight:
263.04
Beilstein:
1976533
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

mp

≥300 °C (lit.)

SMILES string

Oc1ccc(cc1[N+]([O-])=O)[As](O)(O)=O

InChI

1S/C6H6AsNO6/c9-6-2-1-4(7(10,11)12)3-5(6)8(13)14/h1-3,9H,(H2,10,11,12)

InChI key

XMVJITFPVVRMHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service