Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H3410

Sigma-Aldrich

trans-2-Heptene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CHCH3
CAS Number:
Molecular Weight:
98.19
Beilstein:
1719390
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

88 mmHg ( 37.7 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.404 (lit.)

bp

98 °C (lit.)

density

0.701 g/mL at 25 °C (lit.)

SMILES string

CCCC\C=C\C

InChI

1S/C7H14/c1-3-5-7-6-4-2/h3,5H,4,6-7H2,1-2H3/b5-3+

InChI key

OTTZHAVKAVGASB-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shylesh Bhaskaran et al.
Analytical biochemistry, 413(2), 123-132 (2011-03-01)
The nematode Caenorhabditis elegans is a model organism best known for its powerful genetics. There is an increasing need in the worm community to couple genetics with biochemistry. Isolation of functionally active proteins or nucleic acids without the use of
Jaime Onofre et al.
Reproductive biomedicine online, 40(2), 261-269 (2020-02-01)
Which cryopreservation method better protects reproductive potential: the cryopreservation of a testicular cell suspension (TCS) or the cryopreservation of testicular tissue (TET)? Two cryopreservation strategies for spermatogonial stem cells (SSCs) were compared in a mouse model: cryopreservation as TET or
Gestter W L Tessarin et al.
Frontiers in neuroscience, 13, 655-655 (2019-07-19)
Teneurins are type II transmembrane proteins comprised of four phylogenetically conserved homologs (Ten-1-4) that are highly expressed during neurogenesis. An additional bioactive peptide named teneurin C-terminal-associated peptide (TCAP-1-4) is present at the carboxyl terminal of teneurins. The possible correlation between
Enyong Dai et al.
Autophagy, 16(11), 2069-2083 (2020-01-11)
KRAS is the most frequently mutated oncogene in human neoplasia. Despite a large investment to understand the effects of KRAS mutation in cancer cells, the direct effects of the oncogenetic KRAS activation on immune cells remain elusive. Here, we report
Hiroyuki Ohno et al.
Journal of AOAC International, 97(5), 1452-1458 (2015-04-24)
A headspace GC/MS method was developed for identification and quantitation of residual volatile organic compounds in poly(methyl methacrylate) (PMMA) kitchen utensils. A sample was cut into small pieces, then N,N-dimethylacetamide was added in a headspace vial and sealed. After storing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service