Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF97110839

Polycarbonate

(PC), film, thickness 0.5 mm, size 150 × 150 mm, clear

Synonym(s):

CT301350

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$325.00
5 EA
$1,240.00
10 EA
$2,280.00

$325.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$325.00
5 EA
$1,240.00
10 EA
$2,280.00

About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

$325.00


Please contact Customer Service for Availability

Request a Bulk Order

form

film

manufacturer/tradename

Goodfellow 971-108-39

size

150 × 150 mm

thickness

0.5 mm

color

clear

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF38854019GF95515128GF28933241
thickness

0.5 mm

thickness

0.5 mm

thickness

0.5 mm

thickness

0.5 mm

form

film

form

film

form

film

form

film

size

150 × 150 mm

size

300 × 300 mm

size

610 × 915 mm

size

600 × 600 mm

color

clear

color

clear

color

clear

color

-

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service