Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF92143706

Titanium

foil, thickness 0.1 mm, size 100 × 100 mm, purity 99.6+%

Synonym(s):

TI000370

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.6%

form

foil

autoignition temp.

860 °F

manufacturer/tradename

Goodfellow 921-437-06

resistivity

42.0 μΩ-cm, 20°C

size

100 × 100 mm

thickness

0.1 mm

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service