Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF90111068

Calcium

lump, 10 mm max. lump size, weight 100 g, purity 99%

Synonym(s):

CA006101

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$798.00

$798.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$798.00

About This Item

UNSPSC Code:
12141503
NACRES:
NA.23

$798.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99%

form

lumps

manufacturer/tradename

Goodfellow 901-110-68

weight

100 g

max. lump size

10 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service