GF75177153
inconel® 617
rod, Ni57%/Cr22%/Co12%/Mo 9%, 1.6 mm diameter, length 450 mm, temper annealed
Synonym(s):
NE067902
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
rod
manufacturer/tradename
Goodfellow 751-771-53
L
450 mm
diameter
1.6 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Inconel is a registered trademark of Special Metals Corporation
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service