Skip to Content
MilliporeSigma

Skip To

GF67166321

Stainless steel - AISI 304

foil, thickness 0.1 mm, Fe/Cr18%/Ni10%, diameter 25 mm, temper hard

Synonym(s):

FE220271

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 ea
Please contact Customer Service for Availability
$2,290.00

About This Item

NACRES:
NA.23
UNSPSC Code:
11171600

$2,290.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

foil

composition

Fe72Cr18Ni10

reaction suitability

core: chromium, core: iron, core: nickel

manufacturer/tradename

Goodfellow 671-663-21

diam.

25 mm

thickness

0.1 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF62112181GF34069091GF68936529
description

foil, thickness 0.1 mm, Fe/Cr18%/Ni10%, diameter 25 mm, temper hard

description

foil, Fe/Cr18%/Ni10%, thickness 0.1 mm, diameter 10 mm, temper hard

description

foil, Fe/Cr18%/Ni 10%, thickness 0.1 mm, size 150 × 150 mm, temper hard

description

foil, thickness 0.1 mm, Fe/Cr18%/Ni10%, length 1 m, temper hard

form

foil

form

foil

form

foil

form

foil

manufacturer/tradename

Goodfellow 671-663-21

manufacturer/tradename

Goodfellow 621-121-81

manufacturer/tradename

Goodfellow 340-690-91

manufacturer/tradename

Goodfellow 689-365-29

thickness

0.1 mm

thickness

0.1 mm

thickness

0.1 mm

thickness

0.1 mm

reaction suitability

core: iron, core: nickel, core: chromium

reaction suitability

core: chromium, core: iron, core: nickel

reaction suitability

core: iron, core: chromium, core: nickel

reaction suitability

core: chromium, core: nickel, core: iron

diam.

25 mm

diam.

10 mm

diam.

-

diam.

-

composition

Fe72Cr18Ni10

composition

Fe72Cr18Ni10

composition

Fe72Cr18Ni10

composition

Fe72Cr18Ni10


Still not finding the right product?

Explore all of our products under Stainless steel - AISI 304




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters