Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF55589260

Palladium/Nickel

mesh, Pd 95%/Ni 5%, nominal aperture 0.20 mm, size 20 × 20 mm, wire diameter 0.076 mm

Synonym(s):

PJ088710

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

Assay

≥95%

form

mesh

manufacturer/tradename

Goodfellow 555-892-60

nominal aperture

0.20 mm

size

20 × 20 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service