Skip to Content
MilliporeSigma
All Photos(2)

Documents

GF55213657

Platinum

wire reel, 0.1m, diameter 0.175mm, as drawn, 99.95%

Synonym(s):

Platinum, PT005126

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

wire

manufacturer/tradename

Goodfellow 552-136-57

resistivity

10.6 μΩ-cm, 20°C

L × diam.

0.1 m × 0.0175 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Platinum-free catalysts as counter electrodes in dye-sensitized solar cells.
Mingxing Wu et al.
ChemSusChem, 5(8), 1343-1357 (2012-08-14)
A G Quiroga
Journal of inorganic biochemistry, 114, 106-112 (2012-07-24)
This focused review is devoted to our advances in developing trans platinum complexes as antitumoral complexes, in particular those bearing aliphatic amines. It includes some fundamental aspects of the understanding of non conventional metallodrug mechanism. The perspective is arranged by
Xiaoyong Wang et al.
Chemical Society reviews, 42(1), 202-224 (2012-10-09)
Platinum-based anticancer drugs occupy a crucial role in the treatment of various malignant tumours. However, the efficacy and applicability of platinum drugs are heavily restricted by severe systemic toxicities and drug resistance. Different drug targeting and delivery (DTD) strategies have
Xiao-yong Shen et al.
PloS one, 8(8), e69553-e69553 (2013-08-14)
X-ray repair cross-complementing protein 3 (XRCC3) is an essential gene involved in the double-strand break repair pathway. Published evidence has shown controversial results about the relationship between XRCC3 Thr241Met polymorphism and clinical outcomes of non-small cell lung cancer (NSCLC) patients
Josep Ramon Germà-Lluch et al.
Archivos espanoles de urologia, 66(5), 524-535 (2013-06-26)
Cisplatin has been the cornerstone of germ cell testicular tumors therapy since its introduction more tan 30 years ago, and a basic part of the schemes given to multiple ovarian, lung, head and neck, and bladder tumors among others. Some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service