Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF48263014

Silicon carbide

powder, mean particle size 0.1-1 micron, weight 50 g, purity 99%

Synonym(s):

SI516021

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101521
NACRES:
NA.23

Assay

99%

form

powder

manufacturer/tradename

Goodfellow 482-630-14

weight

50 g

mean particle size

0.1-1 μm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service