Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF44891006

Molybdenum

wire, straight, 1000mm, diameter 0.5mm, 99.95%

Synonym(s):

Molybdenum, MO005340

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
MDL number:
UNSPSC Code:
12141727
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

wire

manufacturer/tradename

Goodfellow 448-910-06

resistivity

5.0 μΩ-cm, 20°C

L × diam.

1000 m × 0.5 mm

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad-Reza Rashidi et al.
Expert opinion on drug metabolism & toxicology, 6(2), 133-152 (2010-01-26)
Molybdenum hydroxylases, aldehyde oxidase and xanthine oxidase, are metalloflavoproteins that catalyze both oxidation and reduction of a broad range of drugs and other xenobiotics indicating the importance of these enzymes in drug oxidation, detoxification and activation. Both enzymes are also
Viswanathan S Saji et al.
ChemSusChem, 5(7), 1146-1161 (2012-06-14)
The electrochemical behaviors of molybdenum and its oxides, both in bulk and thin film dimensions, are critical because of their widespread applications in steels, electrocatalysts, electrochromic materials, batteries, sensors, and solar cells. An important area of current interest is electrodeposited
Ralf R Mendel
The Journal of biological chemistry, 288(19), 13165-13172 (2013-03-30)
The transition element molybdenum needs to be complexed by a special cofactor to gain catalytic activity. Molybdenum is bound to a unique pterin, thus forming the molybdenum cofactor (Moco), which, in different variants, is the active compound at the catalytic
Maria Lyra et al.
Hellenic journal of nuclear medicine, 14(1), 49-55 (2011-04-23)
The sleeping giant of molybdenum-99 ((99)Mo) production is grinding to a halt and the world is wondering how this happened. Fewer than 10 reactors in the world are capable of producing radio nuclides for medicine; approximately 50% of the world's
Chantal Iobbi-Nivol et al.
Biochimica et biophysica acta, 1827(8-9), 1086-1101 (2012-12-04)
Molybdenum cofactor (Moco) biosynthesis is an ancient, ubiquitous, and highly conserved pathway leading to the biochemical activation of molybdenum. Moco is the essential component of a group of redox enzymes, which are diverse in terms of their phylogenetic distribution and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service