Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF43979439

Stainless Steel - AISI 304 alloy, FeCr18Ni10

foil, 100 x 100mm, 0.01mm thickness, as rolled, not light tested (nlt)

Synonym(s):

FE220209

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

description

not light tested (nlt)

form

foil (as rolled)

manufacturer/tradename

Goodfellow 439-794-39

size × thickness

100 mm × 100 mm × 0.01 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service