Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF41638282

Aluminum-magnesium alloy, Al92.1Mg7.9

powder, 63micron max. particle size, atomized, 200g

Synonym(s):

AD106010

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352311
NACRES:
NA.23

form

powder (alloy pre-cursor)

manufacturer/tradename

Goodfellow 416-382-82

weight

200 g

max. part. size

63 μm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service