Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF41304899

Aluminum

powder, max. particle size 15 micron, weight 100 g, purity 99%

Synonym(s):

AL006038

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$542.00

$542.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$542.00

About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
MDL number:
UNSPSC Code:
12141702
PubChem Substance ID:
NACRES:
NA.23

$542.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99%

form

powder

autoignition temp.

1400 °F

manufacturer/tradename

Goodfellow 413-048-99

resistivity

2.6548 μΩ-cm

weight

100 g

max. part. size

15 μm

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 6

1 of 6

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service