Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF40542195

Polyethylene - Low Density

(LDPE), sheet, thickness 1 mm, size 150 × 150 mm

Synonym(s):

ET313010

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23
Pricing and availability is not currently available.

form

sheet

manufacturer/tradename

Goodfellow 405-421-95

size

150 × 150 mm

thickness

1 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. GF40542195の購入を検討しておりますが,透明性を教えてください.

    1 answer
    1. This is a product of Goodfellow. The material is not tested for transparancy or transmittance. However the manufacturer suggests the publication below may helpful.
      [Bhuyar, Prakash, et al. "Effect of ultraviolet light on the degradation of low-density and high-density polyethylene characterized by the weight loss and FTIR." Maejo International Journal of Energy and Environmental Communication 1.2 (2019): 26-31.]

      The image below was provided in the above cited paper.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service