Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF37539726

Iron-Chromium-Molybdenum alloy, Fe90Cr9Mo1

foil, 20mm disks, 0.005mm thickness, as rolled

Synonym(s):

FE010220

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

foil (disks - as rolled)

manufacturer/tradename

Goodfellow 375-397-26

size × thickness

20 mm × 0.005 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service