Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF35140586

Ruthenium

powder, max. particle size 450 micron, weight 10 g, purity 99.9%

Synonym(s):

RU006012

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ru
CAS Number:
Molecular Weight:
101.07
MDL number:
UNSPSC Code:
12141739
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

powder

manufacturer/tradename

Goodfellow 351-405-86

resistivity

7.1 μΩ-cm, 0°C

weight

10 g

max. part. size

450 μm

bp

3900 °C (lit.)

mp

2310 °C (lit.)

density

12.45 g/cm3 (lit.)

SMILES string

[Ru]

InChI

1S/Ru

InChI key

KJTLSVCANCCWHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service