Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF30878444

Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3

200mm tube, 8.0mm outside diameter, 1.0mm wall thickness, 6mm inside diameter, annealed

Synonym(s):

FE247500

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 308-784-44

L × wall thickness

200 mm × 1.0 mm

O.D. × I.D.

8.0 mm × 6.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service