Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF26117636

Carbon

rod, 300mm, diameter 25mm, graphite, 99.95%

Synonym(s):

Activated charcoal, C 007950, Charcoal activated

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,320.00

$1,320.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,320.00

About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12141908
PubChem Substance ID:
NACRES:
NA.23

$1,320.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

99.95%

form

rod

autoignition temp.

842 °F

manufacturer/tradename

Goodfellow 261-176-36

resistivity

1375 μΩ-cm, 20°C (graphite)

size diam.

300 mm × 25 mm

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chi-Jung Su et al.
Journal of nanoscience and nanotechnology, 14(6), 4201-4206 (2014-04-18)
Carbon nanofibers (CNFs) have wide applications in energy storage devices, electrically conducting composites, selective adsorbents, and catalyst supports. Catalytic chemical vapor deposition was carried out in this work to synthesize CNFs at mild temperatures of 700 and 800 degrees C.
Cancer treatment: Sharp shooters.
Vivien Marx
Nature, 508(7494), 133-138 (2014-04-04)
Anna Iliadi et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 145(4), 425-433 (2014-04-08)
The aim of this study was to assess the effect of intraoral aging on the setting status of a resin composite and a glass ionomer adhesive, relative to control specimens stored in water. Metallic brackets were bonded with resin composite
Ivar Zekker et al.
Environmental technology, 35(9-12), 1565-1576 (2014-04-08)
Maintaining stability of low concentration (< 1 g L(-1)) floccular biomass in the nitritation-anaerobic ammonium oxidation (anammox) process in the sequencing batch reactor (SBR) system for the treatment of high COD (> 15,000 mg O2 L(-1)) to N (1680 mg
Kimberly T Barrett et al.
Nature, 509(7498), 71-75 (2014-04-22)
One of the most well-recognized stereogenic elements in a chiral molecule is an sp(3)-hybridized carbon atom that is connected to four different substituents. Axes of chirality can also exist about bonds with hindered barriers of rotation; molecules containing such axes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service