Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF25345920

Platinum-tungsten alloy, Pt92W8

5m insulated wire, 0.025mm conductor diameter, 0.007mm insulation thickness, polyimide insulation

Synonym(s):

PT105825

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

description

polyimide insulation

form

wire (insulated)

manufacturer/tradename

Goodfellow 253-459-20

L

5 m

conductor diam.

0.025 mm

insulation thickness

0.007 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service