Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF20882538

Stainless Steel - AISI 302 alloy, FeCr18Ni8

foil, 100 x 100mm, 0.1mm thickness, hard

Synonym(s):

FE200270

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

form

foil (temper hard)

manufacturer/tradename

Goodfellow 208-825-38

size × thickness

100 mm × 100 mm × 0.1 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service