Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF14861583

Polytetrafluoroethylene Coated Glass Fabric

(PTFE 75 / GLASS 25), fabric, weight.m-2-145g, size 500 × 1000 mm, thickness 0.08 mm

Synonym(s):

FP313510

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

form

fabric

manufacturer/tradename

Goodfellow 148-615-83

size

500 × 1000 mm

thickness

0.08 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service