Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF11691468

Stainless Steel - 17-7PH alloy, FeCr17Ni7

foil, 150 x 150mm, 0.05mm thickness, as rolled - condition c, precipitation hardenable, light tight (lt)

Synonym(s):

FE280290

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

description

condition c (precipitation hardenable)
light tight (lt)

form

foil (as rolled
quarter hard)

manufacturer/tradename

Goodfellow 116-914-68

size × thickness

150 mm × 150 mm × 0.05 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service