GF10292688
Aluminum alloy 3003
honeycomb, Al 98%/Mn 1%/Fe 0.5%/Si 0.5%, thickness 20 mm, size 600 × 600 mm, cell size 12.7 mm
Synonym(s):
AB112600
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
honeycomb
manufacturer/tradename
Goodfellow 102-926-88
cell size
12.7 mm
size
600 × 600 mm
thickness
20 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service