Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF09561898

Stainless Steel - 17-7PH alloy, FeCr17Ni7

foil, 300 x 300mm, 0.075mm thickness, annealed - condition a, precipitation hardenable, light tight (lt)

Synonym(s):

FE280300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

description

light tight (lt)

form

annealed (condition a: precipitation hardenable
quarter hard)

manufacturer/tradename

Goodfellow 095-618-98

size × thickness

300 mm × 300 mm × 0.075 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service