Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF06478623

Aluminum-magnesium alloy, Al97Mg3

foil, 0.5m coil, 0.25mm thickness, as rolled

Synonym(s):

AL010260

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352311
NACRES:
NA.23

form

foil (coil - as rolled)

manufacturer/tradename

Goodfellow 064-786-23

size × thickness

0.5 m × 0.25 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service