Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF02532552

Iron

foil, not light tested, 50x50mm, thickness 0.005mm, 99.85%

Synonym(s):

Iron, FE000100

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
UNSPSC Code:
12141721
PubChem Substance ID:
NACRES:
NA.23

Assay

99.85%

form

foil

manufacturer/tradename

Goodfellow 025-325-52

resistivity

9.71 μΩ-cm

L × W × thickness

50 mm × 50 mm × 0.005 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-Won Jang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 59(12), 2503-2507 (2009-06-23)
Zero valent iron has been successfully used for the degradation of a wide range of contaminants. However, this reaction of using ZVI particle produces a large quantity of iron sludge. To solve the problem, we report the synthesis of self-organized
Qiu et al.
Physical review letters, 85(7), 1492-1495 (2000-09-06)
FeF (2) films are grown by the reaction of XeF (2) and SeF (6) with iron foil. The growth initially follows the Mott-Cabrera parabolic rate law, indicating that the process is diffusion limited. At a certain film thickness, however, the
Arnold L Demain et al.
Applied microbiology and biotechnology, 73(1), 55-59 (2006-04-20)
When tetanus toxin is made by fermentation with Clostridium tetani, the traditional source of iron is an insoluble preparation called reduced iron powder. This material removes oxygen from the system by forming FeO(2) (rust). When inoculated in a newly developed
Antonino Missineo et al.
Infection and immunity, 82(6), 2448-2459 (2014-04-02)
Staphylococcus lugdunensis is a coagulase-negative staphylococcus that is a commensal of humans and an opportunistic pathogen. It can cause a spectrum of infections, including those that are associated with the ability to form biofilm, such as occurs with endocarditis or
Maria Wilhelm et al.
Clinical nephrology, 81(4), 251-258 (2014-03-25)
Hyperphosphatemia in advanced chronic kidney disease (CKD) necessitates the use of phosphate binders. This in vitro study assessed phosphate binding and Fe release properties of the novel iron-based phosphate binder PA21. Phosphate adsorption and Fe release were assessed under conditions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service