Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F5687

Sigma-Aldrich

Fmoc-(S)-2-(6-azidohexane)Ala-OH

≥97%

Synonym(s):

(S)-2-{[(9H-fluoren-9-yl)methoxy]carbonylamino}-8-azido-2-methyloctanoic acid, Fmoc-(S)-8-azido-2-methyloctanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H28N4O4
Molecular Weight:
436.50
UNSPSC Code:
12352202
NACRES:
NA.22

Assay

≥97%

reaction suitability

reaction type: click chemistry

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

−20°C

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena M V de Cavanagh et al.
American journal of physiology. Heart and circulatory physiology, 319(4), H744-H752 (2020-08-17)
Patients presenting with classical cardiovascular risk factors within acceptable or average value ranges often develop cardiovascular disease, suggesting that other risk factors need to be considered. Considering that endothelial progenitor cells (EPCs) contribute to endothelial repair, we investigated whether EPCs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service