Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F19906

Sigma-Aldrich

Furfuryl alcohol

98%

Synonym(s):

2-(Hydroxymethyl)furan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10
Beilstein:
106291
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.4 (vs air)

vapor pressure

0.5 mmHg ( 20 °C)
1 mmHg ( 32 °C)
5.5 mmHg ( 55 °C)

Assay

98%

form

liquid

autoignition temp.

915 °F

expl. lim.

16.3 %

refractive index

n20/D 1.486 (lit.)

bp

170 °C (lit.)

mp

−29 °C (lit.)

density

1.135 g/mL at 25 °C (lit.)

SMILES string

OCc1ccco1

InChI

1S/C5H6O2/c6-4-5-2-1-3-7-5/h1-3,6H,4H2

InChI key

XPFVYQJUAUNWIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Nose, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annica Pilgård et al.
Environmental toxicology and chemistry, 29(9), 1918-1924 (2010-09-08)
The furfurylation process is an extensively investigated wood modification process. Furfuryl alcohol molecules penetrate into the wood cell wall and polymerize in situ. This results in a permanent swelling of the wood cell walls. It is unclear whether or not
Marco Minella et al.
Chemosphere, 90(2), 306-311 (2012-08-28)
Suspended particles in a system made up of Aldrich humic acids (HAs) in water account for about 13% of the total HA mass, 10-11% of the organic carbon and 9-11% of radiation extinction in the UVA region. Extinction would be
Rajeev S Assary et al.
Physical chemistry chemical physics : PCCP, 14(48), 16603-16611 (2012-08-31)
Molecular level understanding of acid-catalysed conversion of sugar molecules to platform chemicals such as hydroxy-methyl furfural (HMF), furfuryl alcohol (FAL), and levulinic acid (LA) is essential for efficient biomass conversion. In this paper, the high-level G4MP2 method along with the
Highly selective decarbonylation of 5-(hydroxymethyl)furfural in the presence of compressed carbon dioxide.
Frank M A Geilen et al.
Angewandte Chemie (International ed. in English), 50(30), 6831-6834 (2011-06-11)
Hansruedi Glatt et al.
Mutagenesis, 27(1), 41-48 (2011-08-10)
5-Hydroxymethylfurfural (HMF) and furfuryl alcohol (FFA) are present in numerous foodstuffs at high levels. FFA is also used for the production of polymers. Both compounds had demonstrated some evidence of carcinogenic activity in 2-year bioassays. We tested these compounds and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service