Skip to Content
MilliporeSigma
All Photos(1)

Documents

E31200

Sigma-Aldrich

Ethyl α-hydroxyisobutyrate

98%

Synonym(s):

Ethyl 2-hydroxy-2-methylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(OH)COOC2H5
CAS Number:
Molecular Weight:
132.16
Beilstein:
878308
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.408 (lit.)

bp

150 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C)(C)O

InChI

1S/C6H12O3/c1-4-9-5(7)6(2,3)8/h8H,4H2,1-3H3

InChI key

GFUIDHWFLMPAGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Georgia Lytra et al.
Journal of agricultural and food chemistry, 65(24), 5018-5025 (2017-06-08)
A new method was developed for quantifying substituted acids including, where applicable, their various unexplored enantiomeric forms. A new step was added to acids' usual quantification methods, consisting of extraction, derivatization to methyl esters, and gas chromatography analysis: preliminary extraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service