Skip to Content
MilliporeSigma
All Photos(2)

Documents

E28005

Sigma-Aldrich

Ethyl 2-cyano-3-ethoxyacrylate

98%

Synonym(s):

Ethyl (ethoxymethylene)cyanoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH=C(CN)COOC2H5
CAS Number:
Molecular Weight:
169.18
Beilstein:
878814
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

bp

190-191 °C/30 mmHg (lit.)

mp

49-51 °C (lit.)

SMILES string

CCO\C=C(/C#N)C(=O)OCC

InChI

1S/C8H11NO3/c1-3-11-6-7(5-9)8(10)12-4-2/h6H,3-4H2,1-2H3/b7-6+

InChI key

KTMGNAIGXYODKQ-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service