Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DWKBPM0250P

Azlon Tapered Beakers with Moulded Graduations

polypropylene, capacity 250 mL

Synonym(s):

BPM0250P

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$57.10

$57.10


Please contact Customer Service for Availability


Select a Size

Change View
10 EA
$57.10

About This Item

UNSPSC Code:
41121803
NACRES:
NB.43

$57.10


Please contact Customer Service for Availability

material

polypropylene

packaging

pkg of 10 ea

manufacturer/tradename

Azlon

storage condition

room temperature

assay range

accuracy: 50 mL

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

Application

Excellent solution for measuring and dispensing laboratory solutions and reagents

Features and Benefits

Autoclavable at 121°C and complies with ISO 7056

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service