Skip to Content
MilliporeSigma
All Photos(1)

Documents

DWKBGE230P

Azlon Square Bottles

graduated, cap, Plug, translucent high-density polyethylene, capacity 250 mL

Synonym(s):

BGE230P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121803
NACRES:
NB.43

material

translucent high-density polyethylene

feature

cap Plug
graduated

packaging

pkg of 10 ea

manufacturer/tradename

Azlon

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for storage of liquid and powder samples

Features and Benefits

Space saving square design

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service