Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

DWKBDA212P

Azlon Straight-sided Beakers

graduated, polypropylene, capacity 25 mL

Synonym(s):

BDA212P

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$53.20

$53.20


Please contact Customer Service for Availability


Select a Size

Change View
10 EA
$53.20

About This Item

UNSPSC Code:
41121803
NACRES:
NB.43

$53.20


Please contact Customer Service for Availability

material

polypropylene

feature

graduated

packaging

pkg of 10 ea

manufacturer/tradename

Azlon

storage condition

room temperature

capacity

25 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for general laboratory use and management of liquid samples

Features and Benefits

Straight sided stackable

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service