Key Documents
DWK73664-13
KIMBLE® KIM-KAP® push cap
yellow polypropylene cap, cap diag. 13 mm
Select a Size
1000 EA
$185.00
$185.00
Please contact Customer Service for Availability
Select a Size
Change View
1000 EA
$185.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
$185.00
Please contact Customer Service for Availability
material
yellow polypropylene cap
sterility
non-sterile
packaging
case of 1000 ea
manufacturer/tradename
Kimble® 73664-13
General description
  • KIM-KAP polypropylene closures for culture tubes
  • Autoclavable
  • Color coded
Features and Benefits
  • Autoclavable
  • Color coded
Legal Information
KIM-KAP is a registered trademark of DWK LIFE SCIENCES LLC
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)